accessibility menu, dialog, popup

UserName

NOHOO

CAS RN 100858-33-1

IUPAC Name: benzyl 3-hydroxypyrrolidine-1-carboxylate
Synonyms: benzyl 3-hydroxypyrrolidine-1-carboxylate
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: MBLJFGOKYTZKMH-UHFFFAOYNA-N
SMILES: OC1CCN(C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results