missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-Carbobenzoxy-3-pyrrolidinol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 221.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-1-Carbobenzoxy-3-pyrrolidinol |
Safety and Handling
| TSCA | No |