missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(Boc-amino)-2-propanol, 98%

CAS: 119768-44-4 | C8H17NO3 | 175.23 g/mol

$164.00 - $582.55

Chemical Identifiers

CAS 119768-44-4
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD04974338
InChI Key YNJCFDAODGKHAV-ZCFIWIBFSA-N
Synonym n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
PubChem CID 12094522
IUPAC Name tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
SMILES C[C@@H](O)CNC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2729603
View Documents
Thermo Scientific Chemicals
H2729603
1 g
Each for $164.00
Only null left
 
AAH2729606
View Documents
Thermo Scientific Chemicals
H2729606
5 g
Each for $582.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119768-44-4
175.23
YNJCFDAODGKHAV-ZCFIWIBFSA-N
12094522
C[C@@H](O)CNC(=O)OC(C)(C)C
C8H17NO3
MFCD04974338
n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
tert-butyl N-[(2R)-2-hydroxypropyl]carbamate

Specifications

119768-44-4
1.446
n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
C[C@@H](O)CNC(=O)OC(C)(C)C
1 g
12094522
175.23
(R)-1-(Boc-amino)-2-propanol
C8H17NO3
MFCD04974338
YNJCFDAODGKHAV-ZCFIWIBFSA-N
tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
175.23
−24° (c=1 in Chloroform)
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only