missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(Boc-amino)-2-propanol, 98%

CAS: 119768-44-4 | C8H17NO3 | 175.23 g/mol

Supplier:  Thermo Scientific Chemicals H2729606

 View more versions of this product

Catalog No. AAH2729606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-1-(Boc-amino)-2-propanol
C8H17NO3
MFCD04974338
YNJCFDAODGKHAV-ZCFIWIBFSA-N
tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
175.23
−24° (c=1 in Chloroform)
98%
119768-44-4
1.446
n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
C[C@@H](O)CNC(=O)OC(C)(C)C
5 g
12094522
175.23

Chemical Identifiers

119768-44-4
175.23
YNJCFDAODGKHAV-ZCFIWIBFSA-N
12094522
C[C@@H](O)CNC(=O)OC(C)(C)C
C8H17NO3
MFCD04974338
n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only