missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Benzyl-3-pyrrolidinol 96.0+%, TCI America™

Catalog No. B15325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15325G 5 g
B15321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15325G Supplier TCI America Supplier No. B15325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101930-07-8
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00075484
InChI Key YQMXOIAIYXXXEE-LLVKDONJSA-N
Synonym r-+-1-benzyl-3-pyrrolidinol, r-3-hydroxy-1-benzylpyrrolidine, 3r-1-benzylpyrrolidin-3-ol, r-1-benzyl-3-pyrrolidinol, r-1-benzylpyrrolidin-3-ol, r-1-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-3-hydroxypyrrolidine, r-n-benzyl-3-hydroxypyrrolidine, r-1-benzyl-pyrrolidin-3-ol, r-bhp
PubChem CID 643472
IUPAC Name (3R)-1-benzylpyrrolidin-3-ol
SMILES C1CN(CC1O)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1-Benzyl-3-pyrrolidinol