missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine, 97%, Thermo Scientific Chemicals

$863.23 - $863.23

Chemical Identifiers

CAS 300543-56-0
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.803
MDL Number MFCD11519277
InChI Key UZKBSZSTDQSMDR-QGZVFWFLSA-N
Synonym r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 668697
IUPAC Name 1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6372206
View Documents
Thermo Scientific Chemicals
H6372206
5 g
Each for $863.23
Only null left
 
Description

Description

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

Solubility
Soluble in chloroform, methanol, and dichloromethane. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

300543-56-0
286.803
UZKBSZSTDQSMDR-QGZVFWFLSA-N
668697
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
C17H19ClN2
MFCD11519277
r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine

Specifications

300543-56-0
C17H19ClN2
5 g
Soluble in chloroform,methanol,and dichloromethane. Slightly soluble in water.
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
286.803
286.8
Solid
White
MFCD11519277
r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
UZKBSZSTDQSMDR-QGZVFWFLSA-N
1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
668697
97%
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only