missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6372206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6372206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6372206 Supplier Thermo Scientific Chemicals Supplier No. H6372206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 300543-56-0 | Molecular Formula: C17H19ClN2 | Molecular Weight: 286.803 g/mol

(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine intermediate in the production of (R)-Cetirizine.

Solubility
Soluble in chloroform, methanol, and dichloromethane. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 300543-56-0
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.803
MDL Number MFCD11519277
InChI Key UZKBSZSTDQSMDR-QGZVFWFLSA-N
Synonym r-1-4-chlorophenyl phenyl methyl piperazine, r---1-4-chlorophenyl phenylmethyl piperazine, norchlorcyclizine,-, unii-890a9g29pg, r-1-4-chlorophenyl phenylmethyl piperazine, r-1-p-chlorobenzhydryl piperazine, 1-r-4-chlorophenyl phenyl methyl piperazine, piperazine, 1-r-4-chlorophenyl phenylmethyl, 1-r-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 668697
IUPAC Name 1-[(R)-(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform,methanol,and dichloromethane. Slightly soluble in water.
Formula Weight 286.8
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only