missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.1150g/mL |
| Boiling Point | 232.0°C |
| Quantity | 5 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5410 to 1.5430 |
| Assay Percent Range | 96% min. (GC) |
| Specific Gravity | 1.115 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
RUO – Research Use Only