missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee

Catalog No. AC370650250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC370650250 25 g Glass bottle
AC370650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC370650250 Supplier Thermo Scientific Chemicals Supplier No. 370650250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27298-99-3 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27298-99-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00671639
InChI Key PINPOEWMCLFRRB-ZCFIWIBFSA-N
Synonym r-1-4-chlorophenyl ethylamine, r-1-4-chlorophenyl ethanamine, 1r-1-4-chlorophenyl ethanamine, r-+-1-4-chlorophenyl ethylamine, 1r-1-4-chlorophenyl ethan-1-amine, benzenemethanamine, 4-chloro-alpha-methyl-, alphar, pubchem15222, r-4-chloro-, a-methylbenzylamine, 1r-1-4-chlorophenyl ethylamine, r-1-4-chloro-phenyl ethylamine
PubChem CID 1715226
IUPAC Name (1R)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Color Colorless to Yellow
Density 1.1150g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5410 to 1.5430
Specific Gravity 1.115
Assay Percent Range 96% min. (GC)
Linear Formula ClC6H4CH(CH3)NH2
Boiling Point 232.0°C
Solubility Information Solubility in water: moderately soluble
Formula Weight 155.63
Percent Purity 97% (98% ee)
Chemical Name or Material (R)-1-(4-Chlorophenyl)ethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only