missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Bromophenyl)ethylamine 98.0+%, TCI America™

Catalog No. B36725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36725G 5 g
B36721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36725G Supplier TCI America Supplier No. B36725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 45791-36-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00066025
InChI Key SOZMSEPDYJGBEK-ZCFIWIBFSA-N
Synonym r-1-4-bromophenyl ethylamine, r-+-1-4-bromophenyl ethylamine, r-1-4-bromophenyl ethanamine, 1r-1-4-bromophenyl ethanamine, 1r-1-4-bromophenyl ethan-1-amine, r-1-4-bromophenyl-ethylamine, r-4-bromo-alpha-methylbenzylamine, r-+-4-bromo a-methylbenzylamine, p-bromo-a-methylbenzylamine, pubchem14234
PubChem CID 853000
IUPAC Name (1R)-1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N

Specifications

Melting Point -25°C
Color Yellow
pH 10.5 (4g/L 20°C)
Boiling Point 112°C
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-(4-Bromophenyl)ethylamine
TSCA No
Recommended Storage Refrigerator