missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,3-Butanediol 98.0+%, TCI America™

Catalog No. p-7118886
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11591G 1 g
B11595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11591G Supplier TCI America Supplier No. B11591G
Only null left

Chemical Identifiers

CAS 6290-03-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064277
InChI Key PUPZLCDOIYMWBV-UHFFFAOYNA-N
Synonym r---1,3-butanediol, r-butane-1,3-diol, 3r-butane-1,3-diol, r-1,3-butanediol, 1,3-butanediol, 3r, r---1,3-butylene glycol, r---butane-1,3-diol, 1,3-butanediol, r
PubChem CID 637497
ChEBI CHEBI:52687
IUPAC Name (3R)-butane-1,3-diol
SMILES CC(CCO)O

Specifications

Color Yellow
Boiling Point 117°C
Quantity 1 g
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1,3-Butanediol
RTECSNumber EK0440000
TSCA No