CAS RN 6290-03-5
IUPAC Name:
butane-1,3-diol
Synonyms:
1,3-butanediol butylene glycol 1,3 butylene glycol 1,3-butylene glycol 1,3-butylenglykol methyltrimethylene glycol β-butylene glycol caswell no. 128GG (R)-1,3-butylene glycol (r)-1,3-butylene glycol
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES:
CC(O)CCO
1
–
2
of
2
results
1
Promotions Available
(R)-(-)-1,3-Butanediol, 98%
(R)-(-)-1,3-Butanediol, 98%
CAS: 6290-03-5 MDL Number: MFCD00064277 ChEBI: CHEBI:52687
2
(R)-(-)-1,3-Butanediol 98.0+%, TCI America™
CAS: 6290-03-5 Molecular Formula: C4H10O2 Molecular Weight (g/mol): 90.12 MDL Number: MFCD00064277 InChI Key: PUPZLCDOIYMWBV-UHFFFAOYNA-N Synonym: r---1,3-butanediol,r-butane-1,3-diol,3r-butane-1,3-diol,r-1,3-butanediol,1,3-butanediol, 3r,r---1,3-butylene glycol,r---butane-1,3-diol,1,3-butanediol, r PubChem CID: 637497 ChEBI: CHEBI:52687 IUPAC Name: (3R)-butane-1,3-diol SMILES: CC(CCO)O