missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1,3-Butanediol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 117°C |
| Quantity | 1 g |
| Formula Weight | 90.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (R)-(-)-1,3-Butanediol |
Safety and Handling
| RTECSNumber | EK0440000 |
| TSCA | No |