missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,3-Butanediol, 98%

Catalog No. p-7028014
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2512403 1 g
AAB2512406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2512403 Supplier Thermo Scientific Chemicals Supplier No. B2512403
Only null left

CAS: 6290-03-5 | C8H15NO4 | 189.21 g/mol

(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 6290-03-5
MDL Number MFCD00064277

Specifications

Density 1.005
Boiling Point 203°C to 204°C
Flash Point 121°C (249°F)
Odor Weak
Molecular Formula C8H15NO4
Refractive Index 1.44
Quantity 1 g
Beilstein 1718944
Sensitivity Hygroscopic
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
Solubility Information Fully miscible in water.
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Optical Rotation -30° (Neat)
SMILES CCOC(=O)CNCC(=O)OCC
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol) 189.21
PubChem CID 80502
Formula Weight 90.12
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,3-Butanediol
Show More Show Less
EINECSNumber 228-532-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only