missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98.0+%, TCI America™
$113.59 - $369.64
Chemical Identifiers
| CAS | 127852-28-2 |
|---|---|
| Molecular Formula | C10H8F6O |
| Molecular Weight (g/mol) | 258.16 |
| MDL Number | MFCD03093010 |
| InChI Key | MMSCIQKQJVBPIR-UHFFFAOYNA-N |
| Synonym | r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281 |
| PubChem CID | 2779031 |
| IUPAC Name | 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-ol |
| SMILES | CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Chemical Identifiers
| 127852-28-2 | |
| 258.16 | |
| MMSCIQKQJVBPIR-UHFFFAOYNA-N | |
| 2779031 | |
| CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
| C10H8F6O | |
| MFCD03093010 | |
| r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281 | |
| 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-ol |
Specifications
| 127852-28-2 | |
| White-Yellow | |
| MFCD03093010 | |
| r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281 | |
| CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F | |
| 258.16 | |
| 258.16 | |
| Crystalline Powder |
| 55°C | |
| C10H8F6O | |
| 5 g | |
| MMSCIQKQJVBPIR-UHFFFAOYNA-N | |
| 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-ol | |
| 2779031 | |
| ≥98.0% (GC) | |
| (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol |
Safety and Handling
TSCA : No