missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98.0+%, TCI America™

Catalog No. B41315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41315G 5 g
B413125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41315G Supplier TCI America Supplier No. B41315G
Only null left

Chemical Identifiers

CAS 127852-28-2
Molecular Formula C10H8F6O
Molecular Weight (g/mol) 258.16
MDL Number MFCD03093010
InChI Key MMSCIQKQJVBPIR-UHFFFAOYNA-N
Synonym r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281
PubChem CID 2779031
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-ol
SMILES CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 258.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol