missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

CAS: 103733-65-9 | C10H11NO2 | 177.20 g/mol

Supplier:  Thermo Scientific Chemicals H2911203

 View more versions of this product

Catalog No. AAH2911203

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
C10H11NO2
1 g
BWKMGYQJPOAASG-UHFFFAOYNA-N
OC(=O)C1CC2=CC=CC=C2CN1
177.20
177.2
103733-65-9
MFCD00144038
r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
+163° (c=2 in 1N NaOH)
(3R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
712398

Chemical Identifiers

103733-65-9
177.20
BWKMGYQJPOAASG-UHFFFAOYNA-N
712398
C10H11NO2
MFCD00144038
r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
OC(=O)C1CC2=CC=CC=C2CN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only