accessibility menu, dialog, popup

UserName

HNOHO

CAS RN 103733-65-9

IUPAC Name: 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Synonyms: 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: BWKMGYQJPOAASG-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2=CC=CC=C2CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results