missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Catalog No. AAH29112MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH29112MD 250 mg
AAH2911203 1 g
AAH2911206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH29112MD Supplier Thermo Scientific Chemicals Supplier No. H29112MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103733-65-9 | C10H11NO2 | 177.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103733-65-9
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00144038
InChI Key BWKMGYQJPOAASG-UHFFFAOYNA-N
Synonym r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
PubChem CID 712398
SMILES OC(=O)C1CC2=CC=CC=C2CN1

Specifications

Quantity 250 mg
Optical Rotation +163° (c=2 in 1N NaOH)
IUPAC Name 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Formula Weight 177.2
Chemical Name or Material (R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only