missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Bis(trifluoromethanesulfonate) 96.0+%, TCI America™

Catalog No. b22301g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22301G Supplier TCI America Supplier No. B22301G

Chemical Identifiers

CAS 126613-06-7
Molecular Formula C22H12F6O6S2
Molecular Weight (g/mol) 550.44
MDL Number MFCD00274615
InChI Key OYJLCOSEYYZULE-UHFFFAOYSA-N
Synonym r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
PubChem CID 389883
IUPAC Name 2'-(trifluoromethanesulfonyloxy)-[1,1'-binaphthalen]-2-yl trifluoromethanesulfonate
SMILES FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 550.44
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-1,1′-Binaphthyl-2,2′-diyl Bis(trifluoromethanesulfonate)