accessibility menu, dialog, popup

UserName

FFFSOOOOSOOFFF(?)

CAS RN 126613-06-7

IUPAC Name: 2'-(trifluoromethanesulfonyloxy)-[1,1'-binaphthalen]-2-yl trifluoromethanesulfonate
Synonyms: 2'-(trifluoromethanesulfonyloxy)-[1,1'-binaphthalen]-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 550.44
Molecular Formula: C22H12F6O6S2
InChi Key: OYJLCOSEYYZULE-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)