missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,1'-Binaphthyl-2,2'-disulfonyl Dichloride 98.0+%, TCI America™

Catalog No. p-7123397
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4187200MG 200 mg
B41871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4187200MG Supplier TCI America Supplier No. B4187200MG
Only null left

Chemical Identifiers

CAS 1187629-43-1
Molecular Formula C20H12Cl2O4S2
Molecular Weight (g/mol) 451.33
MDL Number MFCD25372566
InChI Key LPHLPJZENXUTJT-UHFFFAOYSA-N
Synonym (R)-1,1′C-Binaphthyl-2,2′C-disulfonyl Chloride
PubChem CID 46928604
IUPAC Name [1,1'-binaphthalene]-2,2'-disulfonyl dichloride
SMILES ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)S(Cl)(=O)=O

Specifications

Melting Point 235°C
Color White
Quantity 200 mg
UN Number 3261
Formula Weight 451.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1,1′-Binaphthyl-2,2′-disulfonyl Dichloride