missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate), 97%, Thermo Scientific™

Chemical Identifiers

CAS 126613-06-7
Molecular Formula C22H12F6O6S2
Molecular Weight (g/mol) 550.44
MDL Number MFCD00274615
InChI Key OYJLCOSEYYZULE-UHFFFAOYSA-N
Synonym r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
PubChem CID 389883
IUPAC Name [1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
SMILES FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC350432500
View Documents Promotion Details
N/A 250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

126613-06-7
550.44
OYJLCOSEYYZULE-UHFFFAOYSA-N
389883
FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
C22H12F6O6S2
MFCD00274615
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate

Specifications

126613-06-7
100.0
96% min. (HPLC)
(CF3SO3C10H6-)2
- 149.00 (20.00°C c=1,CHCl3)
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
OYJLCOSEYYZULE-UHFFFAOYSA-N
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
389883
97%
96.0
Authentic
C22H12F6O6S2
MFCD00274615
-149
Solubility in water: insoluble
FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
550.44
550.44
(R)-(-)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only