missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1'-Bi-2-naphthol 98.0+%, TCI America™

Catalog No. p-7118876
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11425G 5 g
B114225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11425G Supplier TCI America Supplier No. B11425G
Only null left

Chemical Identifiers

CAS 18531-94-7
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00004068
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 286.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-1,1′-Bi-2-naphthol