missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1'-Bi(2-naphthol), 99%

Catalog No. p-7044805
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0830503 1 g
AAL0830506 5 g
AAL0830514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0830503 Supplier Thermo Scientific Chemicals Supplier No. L0830503
Only null left

CAS: 18531-94-7 | C20H14O2 | 286.33 g/mol

It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

Solubility
Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50 mg/mL.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 18531-94-7
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00004068
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

Melting Point 206°C to 211°C
Density 1.301
Odor Characteristic
Quantity 1 g
UN Number UN2811
Beilstein 3616837
Merck Index 14,1226
Solubility Information Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50mg/mL.
Optical Rotation +34.5 (c=1 in THF)
Formula Weight 286.33
Percent Purity 99%
Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthol)
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DU3106100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only