Learn More
(R)-(+)-1,1'-Bi(2-naphthol), 99%
Description
It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.
Solubility
Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50 mg/mL.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 206°C to 211°C |
| Density | 1.301 |
| Odor | Characteristic |
| Quantity | 1 g |
| UN Number | UN2811 |
| Beilstein | 3616837 |
| Merck Index | 14,1226 |
| Solubility Information | Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50mg/mL. |
| Optical Rotation | +34.5 (c=1 in THF) |
| Formula Weight | 286.33 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H301-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| RTECSNumber | DU3106100 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only