missing translation for 'onlineSavingsMsg'
Learn More

(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals

$476.63 - $859.70

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5H7N3O5
Molecular Weight (g/mol) 189.13
MDL Number MFCD00069337
InChI Key ASNFTDCKZKHJSW-REOHCLBHSA-N
Synonym quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
PubChem CID 40539
ChEBI CHEBI:8734
IUPAC Name (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
SMILES N[C@@H](CN1OC(=O)NC1=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61591LB0
View Documents
Thermo Scientific Chemicals
J61591LB0
5 mg
Each for $476.63
Only null left
 
AAJ61591MA
View Documents
Thermo Scientific Chemicals
J61591MA
10 mg
Each for $859.70
Only null left
 
Description

Description

Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

Solubility
Soluble in ethanol, water. Insoluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

52809-07-1
189.13
ASNFTDCKZKHJSW-REOHCLBHSA-N
40539
(2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
C5H7N3O5
MFCD00069337
quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
CHEBI:8734
N[C@@H](CN1OC(=O)NC1=O)C(O)=O

Specifications

52809-07-1
White
MFCD00069337
1078734
quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
ASNFTDCKZKHJSW-REOHCLBHSA-N
(2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
40539
189.13
Powder
185°C to 187°C
C5H7N3O5
5 mg
14,8085
Soluble in ethanol,water. Insoluble in organic solvents.
N[C@@H](CN1OC(=O)NC1=O)C(O)=O
189.13
CHEBI:8734
≥99%
(+)-Quisqualic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only