missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Quintix™ Pro Balance

Quintix™ Pro laboratory balances are not just about meeting your expectations but exceeding them. They come with an array of premium features that enhance usability, flexibility, and incorporate an eco-friendly design.

Supplier:  Sartorius QTX12201IRO1S

 View more versions of this product

Catalog No. 14-996-904

Only null left
Explore available promotions

Description

Description

  • Enhance the user experience with new premium features
  • Offering a wide range of use in different environments and applications
  • Energy efficiency, reduced carbon footprint, and sustainability, minimizing environmental impact from the manufacturing plant to the bench
Specifications

Specifications

D-sub connector, 9-pin
Color Touchscreen
Real-time Level Support
Quintix Pro Balance
Balance
Class III
12200 g
100 mg
Polybutylene terephthalate (PBT), Glass, Stainless steel
≤0.9s
00 to 240 VAC (± 10%)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions