Learn More
Stoelting™ Quintessential Stereotaxic Injector (QSI)
Program injection parameters (volume, rate and syringe specifications) using the easy-to-use touch screen control box.
$9,550.33 - $12,764.46
Specifications
| For Use With (Application) | Direct injection into the central nervous system using Hamilton microsyringes |
|---|---|
| For Use With (Equipment) | Stereotaxic Instrument |
| Animal | Mouse, Rat |
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|
10000004
|
Stoelting™
53311 |
Injector Head,Touchscreen Control |
Each for $9,550.33
|
|
|||||
|
10000005
|
Stoelting™
53312 |
2 Injector Head,Touchscreen Control |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Control one or two independent injections
- Automatic infusion or withdrawal of nL-μL volumes
- Accommodates Hamilton syringes sized 0.5 to 250 uL
- Available as a single or dual injector
- If purchased as a single injector, add a second injector at any time.
- Preprogrammed syringe specifications
- Out-of-range warnings
Specifications
| Direct injection into the central nervous system using Hamilton microsyringes | |
| Stereotaxic Instrument | |
| Mouse, Rat |