Learn More
Stoelting™ Quintessential Stereotaxic Injector (QSI)
Catalog No. 10000004
Change view
Click to view available options
Includes:
2 Injector Head,Touchscreen Control
Injector Head,Touchscreen Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10000004 Injector Head,Touchscreen Control
10000005 2 Injector Head,Touchscreen Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10000004 Supplier Stoelting™ Supplier No. 53311
Program injection parameters (volume, rate and syringe specifications) using the easy-to-use touch screen control box.
  • Control one or two independent injections
  • Automatic infusion or withdrawal of nL-μL volumes
  • Accommodates Hamilton syringes sized 0.5 to 250 uL
  • Available as a single or dual injector
  • If purchased as a single injector, add a second injector at any time.
  • Preprogrammed syringe specifications
  • Out-of-range warnings
Specifications
For Use With (Application) Direct injection into the central nervous system using Hamilton microsyringes
For Use With (Equipment) Stereotaxic Instrument
Animal Mouse, Rat
Includes Injector Head,Touchscreen Control