missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoxaline-6-boronic acid pinacol ester, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 1167418-13-4 |
|---|---|
| Molecular Formula | C14H17BN2O2 |
| Molecular Weight (g/mol) | 256.11 |
| MDL Number | MFCD11054040 |
| InChI Key | ZYWICCYXTGRUNM-UHFFFAOYSA-N |
| Synonym | 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
| PubChem CID | 44118298 |
| IUPAC Name | 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC=C2N=CC=NC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC445520010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 1167418-13-4 | |
| 256.11 | |
| ZYWICCYXTGRUNM-UHFFFAOYSA-N | |
| 44118298 | |
| CC1(C)OB(OC1(C)C)C1=CC=C2N=CC=NC2=C1 |
| C14H17BN2O2 | |
| MFCD11054040 | |
| 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline |
Specifications
| 1167418-13-4 | |
| 96% min. (GC) | |
| C14H17BN2O2 | |
| 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| CC1(C)OB(OC1(C)C)C1=CC=C2N=CC=NC2=C1 | |
| 256.11 | |
| 256.11 | |
| Quinoxaline-6-boronic acid pinacol ester |
| 130°C to 145°C (0.6mmHg) | |
| Glass Bottle | |
| MFCD11054040 | |
| ZYWICCYXTGRUNM-UHFFFAOYSA-N | |
| 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline | |
| 44118298 | |
| 97% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF IN EYES.
GHS Signal Word: Warning
RUO – Research Use Only