missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoxaline-6-boronic acid pinacol ester, 97%, Thermo Scientific™
Specifications
Specifications
| Boiling Point | 130°C to 145°C (0.6mmHg) |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 256.11 |
| Percent Purity | 97% |
| Chemical Name or Material | Quinoxaline-6-boronic acid pinacol ester |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN. IF IN EYES. |
RUO – Research Use Only