missing translation for 'onlineSavingsMsg'
Learn More

Quinolinium Dichromate 97.0+%, TCI America™

Catalog No. Q005810G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q005810G 10 g
Q005825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q005810G Supplier TCI America Supplier No. Q005810G

Chemical Identifiers

CAS 56549-24-7
Molecular Formula C18H16Cr2N2O7
Molecular Weight (g/mol) 476.325
MDL Number MFCD00059108
InChI Key PYEIMZCGCWSHSV-UHFFFAOYSA-P
Synonym quinolinium dichromate, acmc-209lt3
PubChem CID 11465689
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
SMILES C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 166°C
Quantity 10 g
UN Number 1479
Formula Weight 476.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinolinium Dichromate
TSCA No
Recommended Storage Refrigerator