accessibility menu, dialog, popup

UserName

CAS RN 56549-24-7

IUPAC Name: bis(quinolin-1-ium) [(oxidodioxochromio)oxy]chromiumoylolate
Synonyms: bis(quinolin-1-ium) chromate
Molecular Weight (g/mol): 476.33
Molecular Formula: C18H16Cr2N2O7
InChi Key: PYEIMZCGCWSHSV-UHFFFAOYSA-P
SMILES: [O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O.C1=CC=C2[NH+]=CC=CC2=C1.C1=CC=C2[NH+]=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)