missing translation for 'onlineSavingsMsg'
Learn More

Quinolinium dichromate, 97%, Thermo Scientific™

Catalog No. p-7052685
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5530006 5 g
AAH5530014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5530006 Supplier Thermo Scientific Chemicals Supplier No. H5530006
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 56549-24-7
Molecular Formula C18H16Cr2N2O7
Molecular Weight (g/mol) 476.325
MDL Number MFCD00059108
InChI Key PYEIMZCGCWSHSV-UHFFFAOYSA-P
Synonym quinolinium dichromate, acmc-209lt3
PubChem CID 11465689
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
SMILES C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 164°C to 167°C (decomposition)
Color Orange to Yellow
Quantity 5 g
UN Number UN3087
Formula Weight 476.33
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Quinolinium dichromate
Hazard Category H272-H317-H350
Hazard Statement GHS H Statement
H272-H350-H317
May intensify fire
oxidizer.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures