missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline N-Oxide Hydrate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 59°C |
| Boiling Point | 173°C |
| Quantity | 5 g |
| Formula Weight | 145.16 |
| Percent Purity | ≥97.0% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Quinoline N-Oxide Hydrate |
Safety and Handling
| RTECSNumber | VC2335000 |
| TSCA | No |