accessibility menu, dialog, popup

UserName

NO

CAS RN 198878-42-1

IUPAC Name: quinolin-1-ium-1-olate
Synonyms: quinoline-1-oxide
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: GIIWGCBLYNDKBO-UHFFFAOYSA-N
SMILES: [O-][N+]1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)