missing translation for 'onlineSavingsMsg'
Learn More

Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%

Catalog No. p-7028081
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2520906 5 g
AAB2520914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2520906 Supplier Thermo Scientific Chemicals Supplier No. B2520906
Only null left

CAS: 64201-64-5 | C9H7NO | 145.16 g/mol

Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Notes
Hygroscopic. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 64201-64-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006737
InChI Key GIIWGCBLYNDKBO-UHFFFAOYSA-N
Synonym quinoline n-oxide hydrate, quinoline, 1-oxide, monohydrate, 1-oxidoquinolin-1-ium hydrate, quinoline-n-oxide hydrate, quinoline-1-oxide hydrate, quinolinol, oxamethane, acmc-1be96, quinoline 1-oxide monohydrate, quinoline n-oxide monohydrate
PubChem CID 152363
IUPAC Name 1-oxidoquinolin-1-ium;hydrate
SMILES [O-][N+]1=CC=CC2=CC=CC=C12

Specifications

Melting Point ∼50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 5989589
Sensitivity Hygroscopic
Formula Weight 145.16 (Anhydrous)
Percent Purity 97%
Assay (dry wt.), water ≈20%
Chemical Name or Material Quinoline N-oxide hydrate
EINECSNumber 216-560-6
RTECSNumber VC2340000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only