missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline-8-sulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America Q001625G
Specifications
| Quinoline-8-sulfonyl Chloride | |
| 128°C | |
| 306°C | |
| MFCD00006808 | |
| 3261 | |
| JUYUYCIJACTHMK-UHFFFAOYSA-N | |
| quinoline-8-sulfonyl chloride | |
| 29220 | |
| ≥98.0% (T) |
| 18704-37-5 | |
| White-Yellow | |
| C9H6ClNO2S | |
| 25 g | |
| 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp | |
| C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2 | |
| 227.662 | |
| 227.66 | |
| Crystalline Powder |
Chemical Identifiers
| 18704-37-5 | |
| 227.662 | |
| JUYUYCIJACTHMK-UHFFFAOYSA-N | |
| 29220 | |
| C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2 |
| C9H6ClNO2S | |
| MFCD00006808 | |
| 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp | |
| quinoline-8-sulfonyl chloride |
Safety and Handling
RTECSNumber : VC2830000
TSCA : No