missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-sulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7135620
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00165G 5 g
Q001625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00165G Supplier TCI America Supplier No. Q00165G

Chemical Identifiers

CAS 18704-37-5
Molecular Formula C9H6ClNO2S
Molecular Weight (g/mol) 227.662
MDL Number MFCD00006808
InChI Key JUYUYCIJACTHMK-UHFFFAOYSA-N
Synonym 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp
PubChem CID 29220
IUPAC Name quinoline-8-sulfonyl chloride
SMILES C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2

Specifications

Melting Point 128°C
Color White-Yellow
Boiling Point 306°C
Quantity 5 g
UN Number 3261
Formula Weight 227.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinoline-8-sulfonyl Chloride
RTECSNumber VC2830000
TSCA No