missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-6-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC04501DA
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC04501DA 1 g
CC04501EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC04501DA Supplier Thermo Scientific Chemicals Supplier No. CC04501DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10349-57-2
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00047613
InChI Key VXGYRCVTBHVXMZ-UHFFFAOYSA-N
Synonym 6-quinolinecarboxylic acid, pubchem7578, quinoline-6-carboylic acid, quinolin-6-carboxylic acid, 6-quinoline carboxylic acid, 6-quinoline-carboxylic acid, ksc177e5h, vxgyrcvtbhvxmz-uhfffaoysa, 2-3-cyclopentyl-2-oxo-2,3-dihydro-1h-imidazo 4,5-b pyridin-1-yl-n-2,3-dihydro-1,4-benzodioxin-6-yl acetamide
PubChem CID 82571
IUPAC Name quinoline-6-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)N=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Quinoline-6-carboxylic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.