accessibility menu, dialog, popup

UserName

OHON

CAS RN 10349-57-2

IUPAC Name: quinoline-6-carboxylic acid
Synonyms: 6-quinolinecarboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: VXGYRCVTBHVXMZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)