missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-5-carboxylic acid, 97+%

CAS: 7250-53-5 | C10H7NO2 | 173.17 g/mol

Supplier:  Thermo Scientific Chemicals L1964306

 View more versions of this product

Catalog No. AAL1964306


Only null left
Explore available promotions

Description

Description

Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Quinoline-5-carboxylic acid
>300°C
MFCD00047608
129178
Slightly soluble in water.
OC(=O)C1=C2C=CC=NC2=CC=C1
173.17
173.17
7250-53-5
C10H7NO2
5 g
5-quinolinecarboxylic acid, quinoline-5-carboxylicacid, quinoline-5-carboxylicacid,98+%, zlchem 983, pubchem19952, 5-quinolinecarboxylicacid, acmc-209on5, ksc173e0j, 5-quinolinecarboxylic acid 6ci,7ci,8ci,9ci, quinoline-5-carboxylicacidethylester;5-quinoline-carboxylic acid ethyl ester
RAYMXZBXQCGRGX-UHFFFAOYSA-N
quinoline-5-carboxylic acid
232489
≥97%

Chemical Identifiers

7250-53-5
173.17
RAYMXZBXQCGRGX-UHFFFAOYSA-N
232489
OC(=O)C1=C2C=CC=NC2=CC=C1
C10H7NO2
MFCD00047608
5-quinolinecarboxylic acid, quinoline-5-carboxylicacid, quinoline-5-carboxylicacid,98+%, zlchem 983, pubchem19952, 5-quinolinecarboxylicacid, acmc-209on5, ksc173e0j, 5-quinolinecarboxylic acid 6ci,7ci,8ci,9ci, quinoline-5-carboxylicacidethylester;5-quinoline-carboxylic acid ethyl ester
quinoline-5-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only