missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-5-carboxylic acid, 97+%

Catalog No. p-7047700
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19643MD 250 mg
AAL1964303 1 g
AAL1964306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19643MD Supplier Thermo Scientific Chemicals Supplier No. L19643MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7250-53-5 | C10H7NO2 | 173.17 g/mol

Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-5-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7250-53-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00047608
InChI Key RAYMXZBXQCGRGX-UHFFFAOYSA-N
Synonym 5-quinolinecarboxylic acid, quinoline-5-carboxylicacid, quinoline-5-carboxylicacid,98+%, zlchem 983, pubchem19952, 5-quinolinecarboxylicacid, acmc-209on5, ksc173e0j, 5-quinolinecarboxylic acid 6ci,7ci,8ci,9ci, quinoline-5-carboxylicacidethylester;5-quinoline-carboxylic acid ethyl ester
PubChem CID 232489
IUPAC Name quinoline-5-carboxylic acid
SMILES OC(=O)C1=C2C=CC=NC2=CC=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 129178
Solubility Information Slightly soluble in water.
Formula Weight 173.17
Percent Purity ≥97%
Chemical Name or Material Quinoline-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only