missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-5-carboxaldehyde, 97+%

CAS: 22934-41-4 | C10H7NO | 157.172 g/mol

Supplier:  Thermo Scientific Chemicals L19641MD

 View more versions of this product

Catalog No. AAL19641MD


Only null left
Explore available promotions

Description

Description

Quinoline-5-carboxaldehyde is used as a precursor in the preparation of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-5-carboxaldehyde is used as a precursor in the preparation of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Specifications

Quinoline-5-carboxaldehyde
94°C to 98°C
MFCD00805835
113095
quinoline-5-carboxaldehyde, 5-quinolinecarboxaldehyde, 5-formylquinoline, 5-quinolinecarbaldehyde, acmc-1cmc3, quc
BNEVFKZLYCGDFG-UHFFFAOYSA-N
quinoline-5-carbaldehyde
3543386
≥97%
22934-41-4
C10H7NO
250 mg
Air Sensitive
Slightly soluble in water.
C1=CC(=C2C=CC=NC2=C1)C=O
157.172
157.17

Chemical Identifiers

22934-41-4
157.172
BNEVFKZLYCGDFG-UHFFFAOYSA-N
3543386
C1=CC(=C2C=CC=NC2=C1)C=O
C10H7NO
MFCD00805835
quinoline-5-carboxaldehyde, 5-quinolinecarboxaldehyde, 5-formylquinoline, 5-quinolinecarbaldehyde, acmc-1cmc3, quc
quinoline-5-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only