missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxylic acid, 98+%

Catalog No. p-7046501
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1461103 1 g
AAL1461106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1461103 Supplier Thermo Scientific Chemicals Supplier No. L1461103

CAS: 486-74-8 | C10H7NO2 | 173.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 486-74-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00006782
InChI Key VQMSRUREDGBWKT-UHFFFAOYSA-N
Synonym 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
PubChem CID 10243
ChEBI CHEBI:18311
IUPAC Name quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=N2)C(=O)O

Specifications

Melting Point 253°C to 255°C
Quantity 1 g
Beilstein 5224
Formula Weight 173.17
Percent Purity ≥98%
Chemical Name or Material Quinoline-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-640-1
RTECSNumber GD3850000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only