missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline-4-carboxylic acid, 97%, Thermo Scientific™
$81.93 - $81.93
Chemical Identifiers
| CAS | 486-74-8 |
|---|---|
| Molecular Formula | C10H7NO2 |
| Molecular Weight (g/mol) | 173.171 |
| InChI Key | VQMSRUREDGBWKT-UHFFFAOYSA-N |
| Synonym | 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure |
| PubChem CID | 10243 |
| ChEBI | CHEBI:18311 |
| IUPAC Name | quinoline-4-carboxylic acid |
| SMILES | C1=CC=C2C(=C1)C(=CC=N2)C(=O)O |
Chemical Identifiers
| 486-74-8 | |
| 173.171 | |
| 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure | |
| CHEBI:18311 | |
| C1=CC=C2C(=C1)C(=CC=N2)C(=O)O |
| C10H7NO2 | |
| VQMSRUREDGBWKT-UHFFFAOYSA-N | |
| 10243 | |
| quinoline-4-carboxylic acid |
Specifications
| 486-74-8 | |
| Amber Glass Bottle | |
| 1 g | |
| VQMSRUREDGBWKT-UHFFFAOYSA-N | |
| quinoline-4-carboxylic acid | |
| 10243 | |
| 173.17 | |
| Quinoline-4-carboxylic acid |
| 97% | |
| C10H7NO2 | |
| 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure | |
| C1=CC=C2C(=C1)C(=CC=N2)C(=O)O | |
| 173.171 | |
| CHEBI:18311 | |
| 97% |
Safety and Handling
GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
Warning