missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC04601DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC04601DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC04601DA Supplier Thermo Scientific Chemicals Supplier No. CC04601DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 486-74-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
InChI Key VQMSRUREDGBWKT-UHFFFAOYSA-N
Synonym 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
PubChem CID 10243
ChEBI CHEBI:18311
IUPAC Name quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=N2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Quinoline-4-carboxylic acid
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously