missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$98.79 - $98.79
Chemical Identifiers
| CAS | 191162-39-7 |
|---|---|
| Molecular Formula | C9H8BNO2 |
| Molecular Weight (g/mol) | 172.98 |
| MDL Number | MFCD02183527 |
| InChI Key | YGDICLRMNDWZAK-UHFFFAOYSA-N |
| PubChem CID | 2734663 |
| IUPAC Name | (quinolin-3-yl)boronic acid |
| SMILES | OB(O)C1=CC2=CC=CC=C2N=C1 |
Chemical Identifiers
| 191162-39-7 | |
| 172.98 | |
| YGDICLRMNDWZAK-UHFFFAOYSA-N | |
| (quinolin-3-yl)boronic acid |
| C9H8BNO2 | |
| MFCD02183527 | |
| 2734663 | |
| OB(O)C1=CC2=CC=CC=C2N=C1 |
Specifications
| 191162-39-7 | |
| MFCD02183527 | |
| 3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl | |
| OB(O)C1=CC2=CC=CC=C2N=C1 | |
| 172.98 | |
| 172.98 | |
| Quinoline-3-boronic Acid (contains varying amounts of Anhydride) |
| C9H8BNO2 | |
| 1 g | |
| YGDICLRMNDWZAK-UHFFFAOYSA-N | |
| (quinolin-3-yl)boronic acid | |
| 2734663 | |
| Crystalline Powder |
Safety and Handling
TSCA : No