accessibility menu, dialog, popup

UserName

OHBHON

CAS RN 191162-39-7

IUPAC Name: (quinolin-3-yl)boronic acid
Synonyms: quinolin-3-ylboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: YGDICLRMNDWZAK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CC=C2N=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)